Learn More
Honeywell™ Servus™ Neoprene III™ Boots
Seamless dipped construction keeps this comfortable boot waterproof and pliable in harsh environments
Manufacturer:  Honeywell™ 22114CTM090
 View more versions of this product
Catalog No. 19-051822
Add to cart
Description
Description
Neoprene base shell is dipped twice in neoprene latex, then again in a sealant for triple the protection against caustics, petrochemicals, animal fats, and acids.
  • Fabric lining and Safe-Toe™ steel-toe cap or plain-toe style
  • Chevron sole and heel feature self-cleaning tread pattern with multiple surface-to-surface points of contact for enhanced slip resistance
  • With Breath-O-Prene™ removable, washable insoles
  • Waterproof
Compliance
Meets ASTM™ F2413-11 requirements.
Specifications
Specifications
Safety
12 in.
Neoprene
9