Servus™ Neoprene III™ Boots
Seamless dipped construction keeps this comfortable boot waterproof and pliable in harsh environments
Manufacturer: Honeywell Safety Products 22115-CTM-110
 View all options for this product
Catalog No. 19-160-585
  Check Availability
Add to Cart
    Height (English) 12 in.
    Material Neoprene
    Size 11
    Neoprene base shell is dipped twice in neoprene latex, then again in a sealant for triple the protection against caustics, petrochemicals, animal fats, and acids.
    • Fabric lining and Safe-Toe™ steel-toe cap or plain-toe style
    • Chevron sole and heel feature self-cleaning tread pattern with multiple surface-to-surface points of contact for enhanced slip resistance
    • With Breath-O-Prene™ removable, washable insoles
    • Waterproof
  • Description & Specifications
Related Products
Product Content Feedback