Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Servus™ Dipped Neoprene Safety Boots
Triple-dipped, seamless construction with Chevron outsole
Supplier: Honeywell™ 22114CTM030
This item is not returnable.
View return policy
Description
- Chevron self-cleaning outsole offers a flat, surface-to-surface tread pattern for increased slip resistance, the open channels squeegee liquid and debris out from under foot with each step
- Steel toe construction provides roomy comfort and protection
- Breathe-O-Prene™ removable insoles provide shock absorption and pull moisture away from the feet
- Tough, comfortable fabric lining
- Mid Pac: 12 in. (30.48cm)
- 100% waterproof, noninsulated
- Copper/Tan
- Available in Men's whole sizes 3-15
Compliance
Meets ASTM F2413-11 M I/75 C/75 EH.
Specifications
ANSI Z41 1999 | |
12 in. | |
3 |
Tan | |
Neoprene, Latex |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction