Learn More
Honeywell™ Servus™ PRO™ PVC Knee Boots
Honeywell™ Servus™ PRO™ PVC Knee Boots with Triple Density Technology™ that combines three uniquely formulated compounds, injection-molded to form a rugged, high performance boot with outstanding comfort and durability.
Manufacturer:  Honeywell™ 75102GYM120
 View more versions of this product
Catalog No. 19-121-552H
Add to cart
Description
Description
  • Perimeter of the outsole is formed from a high-density compound that resists abrasion
  • Midsole and interior of the outsole is softer to offer superior grip and comfort
  • A third compound, formulated for flexibility and fit, forms the upper part of the boot; it provides additional abrasion and chemical resistance
  • Seamless construction is 100% waterproof
  • Foot Form™ replaceable contoured cushion insole
  • Steel shank is longer, larger and positioned for optimal support
  • High, wide kick-off lug
  • Self-cleaning triangular cleats
  • Style 75101 boots also feature Safe-Toe™ steel toe cap
Specifications
Specifications
Meets ASTM™ F2413-11 M I/75 C/75
Agriculture, Food Services, Government, Manufacturing, Nuclear, Oil And Gas, Utilities
15 in.
PVC (Polyvinyl Chloride)
12
Knee Boots
Gray
Male
38.1cm
Seamless
TDT™ Dual Compound Yellow And Beige Outsole
Plain Toe