Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Servus PRO PVC Knee Boots
Honeywell Servus PRO PVC Knee Boots with Triple Density Technology that combines three uniquely formulated compounds, injection-molded to form a rugged, high performance boot with outstanding comfort and durability.
$48.25 - $51.75
Specifications
Certifications/Compliance | Meets ASTM™ F2413-11 M I/75 C/75 |
---|---|
For Use With (Application) | Agriculture, Food Services, Government, Manufacturing, Nuclear, Oil And Gas, Utilities |
Material | PVC (Polyvinyl Chloride) |
Seam Type | Seamless |
Sole Material | TDT™ Dual Compound Yellow And Beige Outsole |
Catalog Number | Mfr. No. | Size | Toe Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Toe Material | Price | Quantity & Availability | ||||
19-065-107
|
Honeywell
75101GYM100 |
10 | Steel |
Pair for $51.75
|
|
19-023-900
|
Honeywell
75101GYM140 |
14 |
Pair for $51.75
|
|
19-121-552F
|
Honeywell
75102GYM100 |
10 |
Pair for $48.25
|
|
19-121-552H
|
Honeywell
75102GYM120 |
12 |
Pair for $48.25
|
|
19-039-971
|
Honeywell
75101GYM090 |
9 | Steel |
Pair for $51.75
|
|
Description
- Perimeter of the outsole is formed from a high-density compound that resists abrasion
- Midsole and interior of the outsole is softer to offer superior grip and comfort
- A third compound, formulated for flexibility and fit, forms the upper part of the boot; it provides additional abrasion and chemical resistance
- Seamless construction is 100% waterproof
- Foot Form replaceable contoured cushion insole
- Steel shank is longer, larger and positioned for optimal support
- High, wide kick-off lug
- Self-cleaning triangular cleats
- Style 75101 boots also feature Safe-Toe steel toe cap
Specifications
Meets ASTM™ F2413-11 M I/75 C/75 | |
PVC (Polyvinyl Chloride) | |
TDT™ Dual Compound Yellow And Beige Outsole |
Agriculture, Food Services, Government, Manufacturing, Nuclear, Oil And Gas, Utilities | |
Seamless | |
Knee Boots |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title