Learn More
Honeywell™ Servus™ Pro + Steel-Toe Injection-Molded Hi Boot
Seamless, three-stage PVC injection molded construction keeps feet warm, dry and comfortable
Manufacturer:  Honeywell™ 75105GYM100
 View more versions of this product
Catalog No. 19-120-332F
Add to cart
Description
Description
  • Triple Density Technology (TDT) outsole is engineered with dual compounds, a durable exterior for longer wear and a softer interior to increase surface contact
  • Removable FOOT FORM contour comfort insole provides support for comfort and wicks away moisture
  • Available in gray, US men's sizes: 5 to 7, 9 to 15
  • Hi Boot height is 38.1cm (15 in.)
  • 100 percent waterproof
Compliance
Meets ASTM F2413-11 M I/75 C/75
Specifications
Specifications
ANSI Z41 PT 99
Gray
16 in.
10