Servus™ Rubber Strap-On Overboots

Made from premium lightweight rubber formulated for stretching over bulky work footwear

Manufacturer: Honeywell Safety Products A380YLM110

 View more versions of this product

Catalog No. 19-048166

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Agriculture, construction, food services, utilities
    Height (English) 18 in.
    Liner Material Fabric
    Waterproof Yes
    Material Rubber
    Certifications/Compliance ASTM F2413-11, ANSI Z41-1991, OSHA 1910.136
    Gender Male
    Sole Features Cross Rib™ outsoles add traction
    Color Yellow
    Size 11

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit