Learn More
Honeywell™ Servus™ Rubber Strap-On Overboots
Made from premium lightweight rubber formulated for stretching over bulky work footwear
Manufacturer:  Honeywell™ A380YLM120
 View more versions of this product
Catalog No. 19-048168
Add to cart
Description
Description
  • Special rubber compound prevents ozone deterioration
  • Flexible, hand-layered construction and tough, comfortable fabric lining allow easy on and off
  • Adjustable sidestrap and Cross-Rib™ outsole
  • 18 in. high (46cm)
  • Waterproof
  • Yellow
Specifications
Specifications
Safety
Yellow
18 in.
Rubber
Cross Rib™ outsoles add traction
ASTM F2413-11, ANSI Z41-1991, OSHA 1910.136
Agriculture, construction, food services, utilities
Fabric
12
Yes