Servus™ Rubber Strap-On Overboots

Made from premium lightweight rubber formulated for stretching over bulky work footwear

Manufacturer: Honeywell Safety Products A380YLM130

 View more versions of this product

Catalog No. 19-048169

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-11, ANSI Z41-1991, OSHA 1910.136
    For Use With (Application) Agriculture, construction, food services, utilities
    Gender Male
    Height (English) 18 in.
    Liner Material Fabric
    Sole Features Cross Rib™ outsoles add traction
    Waterproof Yes
    Color Yellow
    Material Rubber
    Size 13

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit