Learn More
Honeywell™ Servus™ SF™ Chem Overboots
Fit over all types of shoes and boots
Manufacturer:  Honeywell™ 11095BLMLRG
 View more versions of this product
Catalog No. 19-122-573B
Add to cart
Description
Description
Provide general chemical resistance for a variety of applications, including WMD and incident response.
  • Super-fit construction
  • Slip-coated interiors
  • Slip-resistant outsoles
  • Seamless, one-piece construction with molded buttons
  • No swelling or softness from exposure to petroleum distillates
  • Height: 12 in. (30.5cm)
  • Made in USA
Compliance
Tested to the chemical permeation requirements of MIL-O-43995C and CID A-A-59520.
Specifications
Specifications
Tested to the chemical permeation requirements of MIL-O-43995C and CID A-A-59520.
Provide general chemical resistance for a variety of applications, including WMD and incident response.
30.5cm
Large, 11 to 13
Black
12 in.
Seamless