Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Servus Studs Second Soles
Steel stud, stretch oversoles fit snugly over shoes and boots
$71.70
Specifications
Color | Black |
---|---|
For Use With (Application) | Flexible for easy storage, heel tab for easy on/off |
Includes | Replaceable Steel studs |
Material | Rubber |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-123-497C
|
Honeywell
SR101XL |
X-Large |
Pair for $71.70
|
|
19-123-497B
|
Honeywell
SR101 |
Large |
Pair for $71.70
|
|
19-123-497D
|
Honeywell
SR101BLM2XL |
2X-Large |
Pair for $71.70
|
|
19-123-497A
|
Honeywell
SR101S/M |
Small |
Pair for $71.70
|
|
Description
Extra-strong stretch rubber construction facilitates snug fit over other footwear and makes for easy storage
- Heel-tab grips for easy on and off
- Replaceablesteel studs
- Black
Specifications
Black | |
Flexible for easy storage, heel tab for easy on/off | |
Replaceable Steel studs | |
Rubber |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title