Servus™ Studs Second Soles

Steel stud, stretch oversoles fit snugly over shoes and boots

Manufacturer: Honeywell Safety Products SR101BLMLRG

 View more versions of this product

Catalog No. 19-123-497B

  Check Availability
Add to Cart

    Specifications

    Product Type Servus Studs™ second soles
    Includes Replaceable Steel studs
    Color Black,Black
    Size Large
    For Use With (Application) Flexible for easy storage, heel tab for easy on/off
    Material Rubber,Rubber

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit