Learn More
Honeywell™ Servus™ XTP™ PVC Non-Safety Toe Work Boots
Injection molded seamless construction for 100% waterproof protection
Manufacturer:  Honeywell™ 75108BLM100
 View more versions of this product
Catalog No. 19-819-118
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Triple Density Technology uses proprietary compound formulations for increased resistance to degradation from many harsh chemicals
  • Dual compound outsole perimeter formulated for tough, long-wearing support and a soft, centersole for better traction and cushion comfort
  • Built-in ergonomic foot bed fights fatigue
  • Higher, wider, kick-off lugs for easy on/off convenience
  • 100% waterproof
  • Height: 15 in. (38.1cm)
  • Black/Red/Gray
Applications:
Ideal for general industrial, farming and occupational use.
Compliance
Meets ASTM™ F2413-11 requirements.
Specifications
Specifications
ANSI Z41 PT 99 I/75; C/75
15 in.
38.1 cm
10
Black
38.1 cm
Seamless