Learn More
Servus™ XTP™ PVC Work Boots
“Xtra Traction Performance” work boots
Manufacturer:  Honeywell Safety Products 75109BLM130
 View more versions of this product
Catalog No. 19-034-465J
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM
Male
Steel
Steel
Black/Yellow/Gray
13
Construction for resistance to organic chemicals found in poultry and fish processing, oil rigs, mining, and general industry
15 in.
Self-cleaning TDT outsole, removable/washable ergonomic insole
Yes
PVC