Servus™ XTP™ PVC Work Boots
“Xtra Traction Performance” work boots
Manufacturer: Honeywell Safety Products 75109BLM150
 View more versions of this product
Catalog No. 19-034-465L
  Check Availability
Add to Cart
    Specifications
    Sole Features Self-cleaning TDT outsole, removable/washable ergonomic insole
    Certifications/Compliance ASTM
    For Use With (Application) Construction for resistance to organic chemicals found in poultry and fish processing, oil rigs, mining, and general industry
    Gender Male
    Height (English) 15 in.
    Shank Material Steel
    Toe Material Steel
    Waterproof Yes
    Color Black/Yellow/Gray
    Material PVC
    Size 15
  • Description & Specifications
Related Products
Product Content Feedback