Servus™ XTP™ PVC Work Boots
“Xtra Traction Performance” work boots
Manufacturer: Honeywell Safety Products 75109BLM060
 View more versions of this product
Catalog No. 19-034-465B
  Check Availability
Add to Cart
    Specifications
    Gender Male
    Height (English) 15 in.
    Shank Material Steel
    Waterproof Yes
    Color Black/Yellow/Gray
    Certifications/Compliance ASTM
    For Use With (Application) Construction for resistance to organic chemicals found in poultry and fish processing, oil rigs, mining, and general industry
    Sole Features Self-cleaning TDT outsole, removable/washable ergonomic insole
    Toe Material Steel
    Material PVC
    Size 6
  • Description & Specifications
Related Products
Product Content Feedback