Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesame Oil, NF, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
8008-74-0
8008-74-0

SE130, 8008-74-0

Spectrum™ Sesame Oil, NF is used as an excipient solvent carrier in suspensions and emulsions, ointments, liniments, ophthalmic preparations, oral capsules and suppositories. All Spectrum Chemical NF are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Water 0.001
Saponification Value 188-195
Quantity 4 L
CAS Max % 1
Grade NF