Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SESN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550756
Description
The antibody detects endogenous levels of total Sestrin-2 / SESN2 protein.
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
Specifications
SESN2 | |
Polyclonal | |
Unconjugated | |
SESN2 | |
Hi95; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; RGD1566319; Ses2; SESN2; SEST2; sestrin 2; sestrin-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P58004 | |
SESN2 | |
Synthetic peptide corresponding to a region derived from internal residues of human sestrin 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction