Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SESN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572834
Description
SESN2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
Specifications
SESN2 | |
Polyclonal | |
Unconjugated | |
SESN2 | |
Hi95; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; RGD1566319; Ses2; SESN2; SEST2; sestrin 2; sestrin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230784, 502988, 83667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P58004, P58043 | |
SESN2 | |
SESTRIN2 antibody for a 19 amino acid peptide near the center of human SESTRIN2. The immunogen is within amino acids 220 - 270 of SESTRIN2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction