Learn More
SESTD1 Mouse anti-Canine, Human, Mouse, Clone: OTI7G4, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA505929
Catalog No. 50-167-6318
Add to cart
Description
Description
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.
Specifications
Specifications
SESTD1
Monoclonal
Unconjugated
SESTD1
Liquid
Sestd1
Full length human recombit protein of human SESTD1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7G4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q80UK0, Q86VW0
SOLO
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
228071, 478823, 91404