Learn More
Sestrin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 213461AP
Catalog No. 50-173-3379
Add to cart
Description
Description
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
Specifications
Specifications
Sestrin 2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P58004, P58043
HI95, SES2, SESN2, SEST2, Sestrin 2, Sestrin2
Rabbit
IgG
150 μL
-20°C
Primary
230784, 83667
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SESN2
Liquid
SESN2
Sestrin 2 Fusion Protein Ag15669
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse