Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SESN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549582

Catalog No. PA549582


Add to cart

Description

Description

Predicted to react with mouse based on sequence homology.

This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.
Specifications

Specifications

SESN2
Polyclonal
PBS with 0.09% sodium azide
P58004
Sestrin-2, Hi95, SESN2, SEST2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83667
Flow Cytometry, Western Blot
Unconjugated
SESN2
Liquid
SESN2
KLH conjugated synthetic peptide between 283-311 amino acids from the Central region of human Sestrin-2.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit