Learn More
SETBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 168411AP
Catalog No. 50-173-1216
Add to cart
Description
Description
This gene encodes a protein which contains a several motifs including a ski homology region and a SET-binding region in addition to three nuclear localization signals. The encoded protein has been shown to bind the SET nuclear oncogene which is involved in DNA replication. Mutations in this gene are associated with Schinzel-Giedion midface retraction syndrome. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
SETBP1
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6X0, Q9Z180
KIAA0437, SEB, SET binding protein, SET binding protein 1, SETBP1
Rabbit
IgG
150 μL
-20°C
Primary
240427, 26040, 291423
Western Blot
Unconjugated
SETBP1
Liquid
SETBP1
SETBP1 Fusion Protein Ag10532
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat