Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SETD1A Recombinant Rabbit Monoclonal Antibody (BLR024E)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | SETD1A |
---|---|
Clone | BLR024E |
Concentration | 10 μg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544243
|
Invitrogen™
MA544243 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544244
|
Invitrogen™
MA544244 |
20 μL |
Each for $173.10
|
|
|||||
Description
SETD1A Recombinant Monoclonal Antibody for Western Blot, IHC (P), IP
SET1A is a component of a histone methyltransferase (HMT) complex that produces mono-, di-, and trimethylated histone H3 at Lys4. The complex is the analog of the S. cerevisiae Set1/COMPASS complex.Specifications
SETD1A | |
10 μg/mL | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
O15047 | |
233904, 9739 | |
Peptide representing a region between residues 1200 and 1250 of human hSET1 (KIAA0339). | |
Antibody |
BLR024E | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
SETD1A | |
BC010250; Histone-lysine N-methyltransferase SETD1A; hSET1A; KIAA0339; KMT2F; Lysine N-methyltransferase 2F; mKIAA0339; mNSC1; non-selective cation channel 1; Nsccn1; SET domain containing 1A; SET domain-containing protein 1A; SET1; Set1/Ash2 histone methyltransferase complex subunit SET1; SET1A; SETD1A | |
SETD1A | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title