Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | SETD4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023305
|
Novus Biologicals
NBP294376100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023306
|
Novus Biologicals
NBP29437620UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
SETD4 Polyclonal antibody specifically detects SETD4 in Human, Mouse samples. It is validated for Western BlotSpecifications
SETD4 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
54093 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
C21orf18, C21orf27, chromosome 21 open reading frame 18, chromosome 21 open reading frame 27, SET domain containing 4, SET domain-containing protein 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 210-300 of human SETD4 (NP_001007260). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title