Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD6 Mouse anti-Human, Clone: GT688, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527851

Catalog No. PIMA527851


Add to cart

Description

Description

Positive Control: K562, THP-1, HL-60 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

SETD9 is a protein-lysine N-methyltransferase. Monomethylates  ft.Lys-310 ft. of the RELA subunit of NF-kappa-B complex, leading to down-regulate NF-kappa-B transcription factor activity. Monomethylates  ft.Lys-8 ft. of H2AZ (H2AZK8me1). Required for the maintece of embryonic stem cell self-renewal.
Specifications

Specifications

SETD6
Monoclonal
2.28 mg/ml
PBS with no preservative
Q8TBK2
SET domain containing 6
Mouse
IgG2b
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
79918
Immunohistochemistry (Paraffin), Western Blot
GT688
Unconjugated
SETD6
Liquid
SETD6
Recombit protein encompassing a sequence within the center region of human SETD6. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit