Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7/9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB10056664
Description
SETD7/9 Polyclonal specifically detects SETD7/9 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SETD7/9 | |
Polyclonal | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Q8WTS6 | |
SETD7 | |
Two synthethic peptides corresponding to amino acids 131-145 (GEVNEDGEMTGEKIA) and 336-352 (GYDHSPPGKSGPEAPEW) of human SETD7 were used as immunogen for this antibody. | |
0.2 mL | |
Histones and Modified Histones, Signal Transduction | |
80854 | |
Human, Mouse | |
Antisera |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay | |
Unconjugated | |
Neat whole antisera with 0.05% Sodium Azide | |
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
This is specific for Set7/9 (a histone H3 lysine 4 methyltransferase) and does not recognize PR-Set7 (a histone H4 lysine 20 methyltransferase). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction