Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SETD7 Rabbit anti-Human, Unconjugated, Clone: ABA-19, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20703
Description
SETD7 Monoclonal antibody specifically detects SETD7 in Human samples. It is validated for IP, WB-Ce.Specifications
SETD7 | |
Monoclonal | |
Unconjugated | |
SETD7 | |
Rabbit | |
100 μg | |
80854 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Western Blot | |
ABA-19 | |
SET domain containing (lysine methyltransferase) 7 | |
FLJ21193, KIAA1717, KMT7, SET7, SET7/9, SET9 | |
A synthetic peptide corresponding to human SETD7 | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction