Learn More
SETD8 Mouse anti-Human, Clone: OTI5E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808217

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Histone methyltransferase that specifically monomethylates ′Lys-20′ of histone H4. H4 ′Lys-20′ monomethylation is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. Mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. Required for cell proliferation, probably by contributing to the maintece of proper higher order structure of DNA during mitosis. Involved in chromosome condensation and proper cytokinesis. Nucleosomes are preferred as substrate compared to free histones.Specifications
SETD8 | |
Monoclonal | |
Unconjugated | |
KMT5A | |
PR-Set7, SET07, SET8, SETD8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
387893 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NQR1 | |
KMT5A | |
Full length human recombit protein of human SETD8 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.