Learn More
SETD8 Rabbit anti-Human, Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191269UV
Catalog No. NBP191269UV
Add to cart
Description
Description
SETD8 Polyclonal antibody specifically detects SETD8 in Human,Mouse samples. It is validated for Western Blot.
Specifications
Specifications
SETD8
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
KMT5A
A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1]
0.1 mL
Primary
387893
Western Blot
Western Blot
H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse