Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Seven2Go™ pH/mV Meter: S2

Ideal for mobile applications in the laboratory, at-line, and outdoors

Manufacturer:  Mettler Toledo™ 30207949

 View more versions of this product

Catalog No. 01-913-965


Add to cart

Description

Description

The Seven2Go portable meters are simple, reliable and are engineered to provide users with fast quality data, one-handed operation and durability.
  • New intuitive menu guarantees out-of-the-box operation for anyone
  • T-Pad provides comfort and improves the speed with which you move through menus
  • Smart button arrangement for faster navigation
  • Automatically recognizes InLab ISM™ (Intelligent Sensor Management) sensor and stores sensor ID, serial number, and last calibration data, thereby reducing chances of error
  • IP67 protection and drop test resistance make these meters weatherproof (withstand harsh and demanding environments)
  • 5-Point calibration (pH)
  • Segmented LCD display
  • Four predefined buffer groups
  • Automatic buffer recognition
  • Linear and segmented calibration methods
  • With user-defined buffer group
  • Ability to perform manual and time-dependent endpoint recording
  • Perform serial measurements in user-defined time interval
  • Visual endpoint signal
  • Automatic/Manual temperature capture
Specifications:
  • pH Measuring Range: -2 to 20
  • pH Resolution: 0.01/0.1
  • pH Accuracy: ±0.01
  • ORP/Redox Measuring Range: -1999 to +1999mV
  • ORP/Redox Resolution: 1
  • ORP/Redox Accuracy: ±1
  • Units: mV, rel. mV
  • Temperature Measuring Range: -5° to +105°C
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • ISM: Light version
  • Memory Size: 200
  • Sensor Inputs: BNC (1012 Ohm)/RCA (cinch)
  • Power Supply: Four 1.5 V AA or 1.3 V NiMH acc.
  • Operating Hours: 250 to 400 hours
  • Dimensions: 222 x 70 x 35mm
  • Weight: 270g
Applications:
  • S2-Standard kit: Wide range of samples including emulsions and suspensions.
  • S2-Field kit: Wide range of samples including emulsions and suspensions; ideal for measurements on the go.
  • S2-Food kit: Solid and semi-solid samples like cheese and meat.
  • S2-Light kit: Academic and educational purposes.
Kits Available:
  • Cat. No. 01-913-965, S2-Meter: Instrument, electrode clip, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-966, S2-Standard kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-967, S2-Field kit: Instrument, electrode clip, InLab™ Expert Pro-ISM-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-968, S2-Food kit: Instrument, electrode clip, InLab™ Solids Pro-IP67, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), bottles for calibration, uGo™ carrying case, operating instructions, quick guide, test report, declaration of conformity and batteries.
  • Cat. No. 01-913-969, S2-Light kit: Instrument, electrode clip, InLab™ Versatile Pro, buffer sachets (pH 4.01, 7.00, 9.21 and 10.00), operating instructions, quick guide, test report, declaration of conformity and batteries.

Certifications

ISO 9001, ISO 14001

Specifications

Specifications

200 (Memory)
0.1°C
270g
±0.01
±0.5°C
4 x AA or 1.3V NiMH acc.
ISO 9001, ISO 14001
0.01 to 0.1
Instrument, electrode clip, operating instructions, quick guide, test report, declaration of conformity and batteries.
BNC (1012 Ohm)/RCA (cinch) (sensor)
1
LCD
-5° to +105°C
Seven2Go S2 pH/mV Meter
±1
250 to 400 hr.
5
Not Applicable
-2 to 20
IP-67
222 x 70 x 35mm
-1999 to +1999 mV
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit