Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ SevenCompact™ Duo S213 pH/Conductivity Meter

Multiparameter measures both pH and conductivity simultaneously or independently. Mettler Toledo SevenCompact Duo213 pH/Conductivity Meter combines precise electrochemical measurement technologies with flexibility, innovative design and ease-of-use.

Manufacturer:  Mettler Toledo™ 30339980

 View more versions of this product

Catalog No. 01-914-327


Add to cart

Includes: Instrument, electrode holder, protective cover, operating instructions, user manual, declaration of conformity, test certificate, InLab™ Pure Pro-ISM for pH and InLab 741-ISM for conductivity, guides to pH and conductivity measurements, two buffer sachets (for pH 4.01, 7.00, 9.21, 10.00), two standard sachets for 1433µS/cm and 12.88mS/cm

Description

Description

  • Robust IP54 design warrants durability
  • Measures TDS, salinity and resistivity
  • Fast and accurate measurements; starting a measurement or calibration, toggling between measurement modes only requires;a single keypress
  • Menu with full sentences in 12 languages without cryptic expressions
  • User-friendly operation
  • Clear, well arranged color display
  • Many peripheral options
  • GLP compliant
  • Service offering, including IQ/OQ
  • Option to connect various accessories such as barcode reader, keyboard, USB stick, USB printer or a PC with EasyDirect™ pH software
  • Workflow efficiency can be increased with the uPlace sensor stand and uMix stirrer
Specifications
  • pH Range: -2.000 to 20.000
  • pH Resolution: 0.001, 0.01, 0.1
  • pH Accuracy: ±0.002
  • mV Range: -2000.0 to 2000.0
  • mV Resolution: 0.1, 1
  • mV Accuracy: ±0.1mV in -1000 to 1000 mV; ±0.2mV outside
  • Relative mV Range: -2000.0 to 2000.0
  • Relative mV Resolution: 0.1, 1
  • Relative mV Accuracy: ±0.1mV in -1000 to 1000 mV; ±0.2mV outside
  • Conductivity Range: 0.001µS/cm to 1000mS/cm
  • Conductivity Resolution: 0.001 to 1
  • Conductivity Accuracy: ±0.5%
  • TDS Range: 0.00mg/L to 1000g/L
  • TDS Resolution: 0.01 to 1
  • TDS Accuracy: ±0.5%
  • Salinity Range: 0.00 to 80.00 psu/ppt
  • Salinity Resolution: 0.00 to 1.0
  • Salinity Accuracy: ±0.5%
  • Resistivity Range: 0.00 to 100.0 MOhm-cm
  • Resistivity Resolution: 0.00 to 1.0
  • Resistivity Accuracy: ±0.5%
  • Conductivity ash Range: 0.000 to 2022%
  • Conductivity ash Resolution: 0.001 to 1
  • Conductivity ash Accuracy: ±0.5%
  • Temperature Range: -5° to +130°C (ATC); -30° to +130°C (MTC)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ± 0.1°C
Calibration
  • Calibration Points: Conductivity 1; pH Max. 5
  • Cond. Standards and Buffer groups: Eight pre-defined pH buffers and 13 pre-defined cond. STD
  • Buffer Recognition: One user defined each
  • Isopotential Point: pH 7.00
  • Temperature Capture: Automatic and manual
  • Calibration Reminder: User-configurable
  • Electrode Status Icon: Shows quality of last calibration
  • Manual Input of Cell Constant: Yes
  • Display of Cell Constant: In conductivity and dual channel mode
Measurement
  • Choice of Endpoint Formats: Automatic, manual, timed
  • Choice of Stability Criteria (pH only): Fast, standard, strict
  • Choice of pH Decimal Places: 1.111, 1.11, 1.1
  • Serial Measurements: Time interval, user-configurable
  • Choice of Reference Temperature: 20° or 25°C
  • Choice of Temperature Compensation: Linear, non-linear, off, pure-water
System and GLP
  • Display Color Scheme: Blue, red, green, grey
  • Time and Date: Two time formats, four date formats
  • PIN Protection: Login, deletion of data, system settings
  • Measurement Limits: User-defined
  • Instrument Modes: Routine mode/expert mode
  • ID Input: User ID, sample ID, sensor ID, sensor SN
  • ID Input Automation: Barcode reader and keyboard (USB)
Data Processing
  • Data Storage: 2000 measurements
  • Print Formats: Short, normal, GLP
  • Data Export: USB-stick, EasyDirect pH PC software
Connectors
  • pH Sensor Input: BNC (impedance >3 x 1012 Ohm)
  • Temperature Sensor Input: Cinch/RCA (NTC30 kOhm or Pt1000)
  • Reference Input: 2mm Ref.
  • Digital sensor Input: Mini-LTW
  • Conductivity Input: Mini-DIN
  • Conductivity Input: RS232, USB-A, USA-B
  • Stirrer Connector: Mini-DIN
General
  • Display: Color TFT
  • Power requirements: DC 9 to 12V, 2.5w
  • Dimensions: 204 x 174 x 74mm
  • Weight: 890g
Ambient Conditions
  • Temperature: 5° to 40°C
  • Relative humidity: 5 to 80% (non-condensing)
  • Housing: ABS/PC reinforced
  • Window: Polymethyl methacrylate (PMMA)
  • Keypad: Membrane keypad: Polyethylene terephthalate (PET)
Regulations
  • Pure and ultra-pure water: USP/EP compliant
  • Bioethanol: ABNT NBR 10547:2006
  • Sugar analysis: ICUMSA GS2/3-17 and GS 1/3/4/7/8-13
Specifications

Specifications

Instrument, electrode holder, protective cover, operating instructions, user manual, declaration of conformity, test certificate, InLab™ Pure Pro-ISM for pH and InLab 741-ISM for conductivity, guides to pH and conductivity measurements, two buffer sachets (for pH 4.01, 7.00, 9.21, 10.00), two standard sachets for 1433μS/cm and 12.88mS/cm
0.001, 0.01, 0.1
-2000.0 to 2000.0
0.1, 1
pH 7.00
0.001µS/cm to 1000mS/cm
0.00mg/L to 1000g/L
±0.5%
0.00 to 1.0
0.00 to 100.0 MOhm-cm
±0.5%
Max. 5
1
0.1°C
Automatic or manual
Membrane keypad: Polyethylene terephthalate (PET)
USB-stick, EasyDirect pH PC software
12 languages
IP54
20.4 x 17.4 x 7.4cm
SevenCompact Duo S213
-2.000 to 20.000
±0.002
-2000.0 to 2000.0
±0.1mV in -1000 to 1000 mV; ±0.2mV outside
0.001 to 1
±0.5%
0.01 to 1
0.00 to 80.00 psu/ppt
±0.5%
0.00 to 1.0
Linear, non-linear, off, pure-water
Conductivity 1; pH Max. 5
-5° to +130°C (ATC); -30° to +130°C (MTC)
± 0.1°C
Color TFT
Routine mode/expert mode
2000 measurements
9 to 12V DC, 2.5w
5 to 80% (non-condensing)
890g
S213-Water kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit