Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEZ6L2/BSRP-A Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297264APC
Description
SEZ6L2/BSRP-A Polyclonal antibody specifically detects SEZ6L2/BSRP-A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SEZ6L2/BSRP-A | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
seizure 6-like protein 2, seizure related 6 homolog (mouse)-like 2, UNQ1903/PRO4349 | |
Produced in rabbits immunized with E. coli-derived Human SEZ6L2/BSRP-A fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
26470 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction