Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Mouse anti-Human, Clone: NR5A1/3397, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30780610UG
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
SF-1/NR5A1/Steroidogenic Factor 1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
Recombinant fragment (around aa220-461) of human SF-1/NR5A1/Steroidogenic Factor 1 protein | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Peptide Array | |
NR5A1/3397 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
2516 | |
Store at 4°C. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title