Learn More
Invitrogen™ SF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592965
Description
Immunogen sequence: MATGANATPL DFPSKKRKRS RWNQDTMEQK TVIPGMPTVI PPGLTREQER AYIVQLQIED LTRKLRTGDL GIPPNPEDRS PSPEPIYNSE GKRLNTREFR TRKKLEEERH NLITEMVALN PDFKPPADYK PPATRVSDKV MIPQDEYPEI NFVGLLIGPR GNTLKNIEKE CNAKIMIRGK GSVKEGKVGR KDGQMLPGED EPLHALVTAN TMENVKKAVE QIRNILKQGI ETPEDQNDLR KMQLRELARL NGTLREDDNR ILRPWQSSET RSITNTTVCT; Positive Samples: HepG2; Cellular Location: Nucleus.
Splicing factor 1 is a nuclear pre-mRNA splicing factor. The protein specifically recognizes the intron branch point sequence at the 3' splice site, together with the large subunit of U2 auxiliary factor (U2AF), and is required for the early stages of spliceosome assembly. It also plays a role in nuclear pre-mRNA retention and transcriptional repression. The protein contains an N-terminal U2AF ligand motif, a central hnRNP K homology motif and quaking 2 region which bind a key branch-site adenosine within the branch point sequence, a zinc knuckles domain, and a C-terminal proline-rich domain. Alternative splicing results in multiple transcript variants.
Specifications
SF1 | |
Polyclonal | |
Unconjugated | |
SF1 | |
BBP; D11S636; M ZFM1; Mammalian branch point-binding protein; MBBP; MZFM; SF; SF1; Splicing factor 1; transcription factor ZFM1; WBP4; wu:fc09f06; ZCCHC25; ZFM1; Zfp162; Zinc finger gene in MEN1 locus; zinc finger protein 162; ZNF162 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22668, 7536 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.06 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15637, Q64213 | |
SF1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human SF1 (NP_9737271). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.