Learn More
Selleck Chemical LLCSupplier Diversity Partner SF2523 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85892MG
SF2523 is a highly selective and potent inhibitor of PI3K with IC50 values of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.
Catalog No. 50-194-8196