Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B5 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259132

Catalog No. NBP259132


Add to cart

Description

Description

SF3B5 Polyclonal antibody specifically detects SF3B5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

SF3B5
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
MGC3133, Pre-mRNA-splicing factor SF3b 10 kDa subunit, SF3b10, SF3b5, splicing factor 3B subunit 5, splicing factor 3b, subunit 5, 10kDa, Ysf3
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human, Mouse
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
SF3B5
Maps to a region between 1 and 50
Immunogen affinity purified
RUO
Primary
83443
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only