Learn More
SF3B5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155251AP
Catalog No. 50-173-0287
Add to cart
Description
Description
SF3B5 is a protein coding gene involved in pre-mRNA splicing as a component of the splicing factor SF3B complex, a constituent of the spliceosome. SF3B complex is required for  ft.A ft. complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA.
Specifications
Specifications
SF3B5
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q923D4, Q9BWJ5
SF3b10, SF3B5, Splicing factor 3B subunit 5
Rabbit
IgG
150 μL
-20°C
Primary
66125, 680891, 83443
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SF3B5
Liquid
Sf3b5
SF3B5 Fusion Protein Ag7205
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat