Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571316
Description
This target displays homology in the following species: Dog: 83%; Human: 100%.
SFI1 gene ontology annotations related to this gene include negative regulation of phosphatase activity.
Specifications
SFI1 | |
Polyclonal | |
Unconjugated | |
Sfi1 | |
homolog of yeast Sfi1; hSFI1; hSfi1p; KIAA0542; PISD; PPP1R139; protein phosphatase 1, regulatory subunit 139; protein SFI1 homolog; Sfi1; SFI1 centrin binding protein; Sfi1 homolog, spindle assembly associated; Sfi1 homolog, spindle assembly associated (yeast) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9814 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A8K8P3 | |
Sfi1 | |
synthetic peptide directed towards the C-terminal region of human SFI1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction