Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sFRP-1 Rabbit anti-Human, Clone: 24, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289596100UL
Description
sFRP-1 Monoclonal specifically detects sFRP-1 in Human samples. It is validated for ELISA.Specifications
sFRP-1 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
24 | |
ELISA 1:5000-1:10000 | |
FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
Recombinant Human sFRP-1 Protein | |
100 μg | |
Apoptosis | |
6422 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction