Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFRP1 Recombinant Rabbit Monoclonal Antibody (5I6V1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538193
Description
Positive Samples: U-251MG, Mouse kidney, Rat testis, Rat kidney Immunogen sequence: MGIGRSEGGR RGAALGVLLA LGAALLAVGS ASEYDYVSFQ SDIGPYQSGR FYTKPPQCVD IPADLRLCHN VGYKKMVLPN LLEHETMAEV KQQASSWVPL.
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.
Specifications
SFRP1 | |
Recombinant Monoclonal | |
0.88 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C4U3, Q8N474, Q9R168 | |
SFRP1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SFRP1 (Q8N474). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
5I6V1 | |
Unconjugated | |
SFRP1 | |
2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20377, 6422, 84402 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction