Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRP2 Polyclonal Antibody
SFRP2 Polyclonal Antibody
Supplier: Thermo Scientific PA579985
Description
SFRP2 Polyclonal Antibody for Western Blot, ELISA

Specifications
SFRP2 | |
Polyclonal | |
Unconjugated | |
SFRP2 | |
AI851596; FKSG12; FRP 2; FRP2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP1; SARP-1; SDF 5; Sdf5; SDF-5; secreted apoptosis related protein 1; secreted apoptosis-related protein 1; secreted frizzled related protein 2; secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; SFRP2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170; UNQ361/PRO697 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6423 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q96HF1 | |
SFRP2 | |
E.coli-derived human SFRP2 recombinant protein (Position: L104-C295). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction