Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS1 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for SFRS1

Manufacturer:  Abnova Corporation H00006426T01

Catalog No. 89-002-048


Add to cart

Description

Description

This gene encodes a member of the arginine/serine-rich splicing factor protein family, and functions in both constitutive and alternative pre-mRNA splicing. The protein binds to pre-mRNA transcripts and components of the spliceosome, and can either activate or repress splicing depending on the location of the pre-mRNA binding site.

  • Splicing factor, arginine/serine-rich 1
  • Transfected cell line: 293T
  • Plasmid: pCMV-SFRS1 full-length
  • Host: Human
  • Theoretical molecular weight: 27.39kDa
  • Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)

Western Blotting

Specifications

Specifications

Western Blot
293T
BC010264
6.8
Transient overexpression cell Lysate was tested with Anti-SFRS1 antibody (H00006426-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
SFRS1 293T Cell Transient Overexpression Lysate(Denatured)
27.39
pCMV-SFRS1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit