Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFRS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541787
Description
Peptide sequence: MSDSGEQNYG ERESRSASRS GSAHGSGKSA RHTPARSRSK EDSRRSRSKS Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.
Specifications
SFRS10 | |
Polyclonal | |
Unconjugated | |
TRA2B | |
5730405G21Rik; D16Ertd266e; Htra2-beta; PPP1R156; protein phosphatase 1, regulatory subunit 156; RA301; serine/arginine-rich splicing factor 10; SFRS10; SIG-41; Silg41; Silica-induced gene 41 protein; similar to splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich 10; splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila); splicing factor, arginine/serine-rich, 10; SRFS10; Srsf10; TRA-2 beta; Tra2b; TRA2beta; TRA2-beta; TRAN2B; transformer 2 beta homolog; transformer 2 beta homolog (Drosophila); transformer-2 protein homolog B; Transformer-2 protein homolog beta; zgc:56612 | |
Rabbit | |
Protein A | |
RUO | |
6434 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P62995 | |
TRA2B | |
Synthetic peptide directed towards the N-terminal of human SFRS10 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction