Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFRS15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555982
Description
Immunogen sequence: KVKPEELESF CEGGMLDSDT LNPDWKGIPK KPENEVAQNG GAETSHTEPV SPIPKPLPVP VPPIPVPAPI TVPPPQVPPH QPGPPVVGAL QP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
The family of SR factors all contain one or more RNA recognition motifs (RRM) and an SR-rich domain. They are not only essential for constitutive splicing, but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. Splicing factor arginine/serine-rich 15 (SFRS15), also designated CTD-binding SR-like protein RA4, contains one RRM and one SR-rich domains. SFRS15 interacts with C-terminal repetitive domain (CTD) of Pol II and is believed to functionally and physically link transcription and pre-mRNA processing. Localized to the nucleus, SFRS15 is expressed as two isoforms produced by alternative splicing.
Specifications
SFRS15 | |
Polyclonal | |
Unconjugated | |
SCAF4 | |
AA517739; CTD-binding SR-like protein RA4; KIAA1172; pre-mRNA splicing SR protein rA4; SCAF4; serine/arginine-rich splicing factor 15; SFR15; SFRS15; splicing factor serine alanine 15; splicing factor, arginine/serine-rich 15; SRA4; SR-like CTD-associated factor 4; SR-related and CTD-associated factor 4; SR-related CTD associated factor 4; SR-related CTD-associated factor 4; Srsf15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
224432, 57466 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95104 | |
SCAF4 | |
Recombinant protein corresponding to Human SFRS15. Recombinant protein control fragment (Product #RP-95879). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction