Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS18 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079851C
Description
SFRS18 Polyclonal antibody specifically detects SFRS18 in Human samples. It is validated for Western BlotSpecifications
SFRS18 | |
Polyclonal | |
Western Blot | |
bA98I9.2, C6orf111, chromosome 6 open reading frame 111, DKFZp564B0769, FLJ14752, FLJ14992, FLJ90147, HSPC306, MGC104269, RP11-98I9.2, Serine/arginine-rich-splicing regulatory protein 130, serine-arginine-rich-splicing regulatory protein 130, Splicing factor, arginine/serine-rich 130, splicing factor, arginine/serine-rich 18, SR rich protein, SR-related protein, SR-rich protein, SRRP130, SRrp130FLJ14853 | |
The immunogen recognized by this antibody maps to a region between residue 200 to 250 of human splicing factor, arginine/serine-rich 18 (splicing factor, arginine/serine-rich 130) using the numbering given in entry NP_056306.1 (GeneID 25957). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
25957 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction