Learn More
SFRS2IP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | SFRS2IP |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2278
![]() |
Bethyl Laboratories, Inc
IHC00541T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2277
![]() |
Bethyl Laboratories, Inc
IHC00541 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Dog, Rabbit, Orangutan, Monkey, Gorilla, Chimpanzee, Little brown bat, hirteen-lined ground squir
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.Specifications
SFRS2IP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CASP11, CTD-associated SR protein 11, renal carcinoma antigen NY-REN-40, SC35-interacting protein 1, serine/arginine-rich splicing factor 2, interacting protein, serine/arginine-rich splicing factor 2-interacting protein, SFRS2-interacting protein, SFRS2IP, SIP1, splicing factor, arginine/serine-rich 2, interacting protein, splicing factor, arginine/serine-rich 2-interacting protein, splicing regulatory protein 129, SR-related and CTD-associated factor 11, SR-related CTD-associated factor 11, SRRP129, SRSF2-interacting protein, SRSF2IP | |
SCAF11 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q99590 | |
9169 | |
Between 1098 and C-term | |
Primary | |
4° C | |
SCAF11 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.