Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SFRS3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157242
|
Novus Biologicals
NBP157242 |
100 μL |
Each of 1 for $436.00
|
|
Description
SFRS3 Polyclonal specifically detects SFRS3 in Human samples. It is validated for Western Blot.Specifications
SFRS3 | |
Polyclonal | |
Rabbit | |
P84103 | |
6428 | |
Synthetic peptides corresponding to SFRS3(splicing factor, arginine/serine-rich 3) The peptide sequence was selected from the N terminal of SFRS3. Peptide sequence SVWVARNPPGFAFVEFEDPRDAADAVRELDGRTLCGCRVRVELSNGEKRS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
Pre-mRNA-splicing factor SRP20, serine/arginine-rich splicing factor 3, SFRS3splicing factor, arginine/serine-rich, 20-kD, Splicing factor, arginine/serine-rich 3SRP20, SRp20 | |
SRSF3 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title