Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157466
Description
SFRS8 Polyclonal specifically detects SFRS8 in Human samples. It is validated for Western Blot.Specifications
SFRS8 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
B2RN45 | |
SFSWAP | |
Synthetic peptides corresponding to SFRS8(splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila)) The peptide sequence was selected from the N terminal of SFRS8. Peptide sequence LVFGYACKLFRDDERALAQEQGQHLIPWMGDHKILIDRYDGRGHLHDLSE The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
Drosophila homolog), Drosophila), SFRS8, Splicing factor, arginine/serine-rich 8, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, splicing factor, suppressor of white-apricot homolog, splicing factor, suppressor of white-apricot homolog (Drosophila), Suppressor of white apricot protein homolog, SWAPMGC167082 | |
Rabbit | |
Affinity Purified | |
RUO | |
6433 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title