Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SFTPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115987
Description
Antibody detects endogenous levels of total Surfactant Protein A.
This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication.
Specifications
SFTPA2 | |
Polyclonal | |
Unconjugated | |
SFTPA2 | |
35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC5; collectin 5; Collectin-5; PSAP; PSPA; PSP-A; Pulmonary surfactant-associated protein A2; SFTP1; SFTPA; SFTPA2; SFTPA2B; SP-2A; SP-2A beta; SP-2A gamma; SP-A; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant protein A2; surfactant, pulmonary-associated protein A2A | |
Rabbit | |
Affinity chromatography | |
RUO | |
729238 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q8IWL1 | |
SFTPA2 | |
A synthesized peptide derived from human SFTPA2(Accession Q8IWL1), corresponding to amino acid residues G43-E93. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction